Skip to Content
Merck

O115

Oxymetholone

Synonym(s):

17β-Hydroxy-2-(hydroxymethylene)-17-methyl-5α-androstan-3-one, Anasterone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
EC Number:
207-098-6
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

[H][C@@]12CC[C@]3([H])C(CC[C@@]4(C)[C@@]3([H])CC[C@]4(C)O)[C@@]1(C)C\C(=C\O)C(=O)C2

InChI

1S/C21H32O3/c1-19-11-13(12-22)18(23)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,22,24H,4-11H2,1-3H3/b13-12-/t14-,15+,16?,17-,19-,20-,21-/m0/s1

InChI key

ICMWWNHDUZJFDW-ZDBQEZQZSA-N



Still not finding the right product?

Explore all of our products under Oxymetholone


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library