Skip to Content
Merck
All Photos(1)

Key Documents

N6162

Sigma-Aldrich

Anti-Neuropeptide FF1 Receptor (NPFF1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-G protein-coupled Receptor 147 (GPR147), Anti-RFamide-related Peptide Receptor OT7T022

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:1,000 using brain lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPFFR1(64106)

General description

Neuropeptide FF receptor 1 (NPFF1) is a receptor for the protein RFamide-related peptide-3 (RFRP-3). It is expressed in the ovaries and the gene encoding it is localized on human chromosome 10q21.

Immunogen

peptide corresponding to the C-terminal sequence (amino acids 418-430) and N-terminal sequence (amino acids 1-16) of the NPFF1 protein.

Biochem/physiol Actions

Neuropeptide FF receptor 1 (NPFF1) along with its ligand, prevents gonadotropin signaling.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and characterization of two G protein-coupled receptors for neuropeptide FF.
Bonini JA
The Journal of Biological Chemistry, 275(50), 39324-39331 (2000)
Hajime Oishi et al.
Endocrinology, 153(7), 3435-3445 (2012-06-14)
RFamide-related peptide-3 (RFRP-3), a mammalian ortholog of avian gonadotropin-inhibitory hormone, has pronounced inhibitory effects on reproduction in a number of species. RFRP-3 suppresses gonadotropin release at the hypothalamic and/or pituitary levels; however, increasing evidence also suggests putative functions within the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service