Skip to Content
Merck

N3535

(S)-Nafadotride tartrate

>98% (HPLC), solid

Synonym(s):

(−)-N[n-Butyl-2-pyrrolidinyl)methyl]-1-methoxy-4-cyanonaphthalene-2-carboxamide tartrate, 2-Naphthalenecarboxamide tartrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H33O8N3
CAS Number:
Molecular Weight:
515.56
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>98% (HPLC)

form

solid

color

white

solubility

H2O: soluble ~9 mg/mL

storage temp.

2-8°C

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O.[H][C@]1(CCCN1CCCC)CNC(=O)c2cc(C#N)c3ccccc3c2OC

InChI

1S/C22H27N3O2.C4H6O6/c1-3-4-11-25-12-7-8-17(25)15-24-22(26)20-13-16(14-23)18-9-5-6-10-19(18)21(20)27-2;5-1(3(7)8)2(6)4(9)10/h5-6,9-10,13,17H,3-4,7-8,11-12,15H2,1-2H3,(H,24,26);1-2,5-6H,(H,7,8)(H,9,10)/t17-;1-,2-/m00/s1

InChI key

WOPHJTZRFZTRKU-FJMPKIMZSA-N

Biochem/physiol Actions

D3 Dopamine receptor antagonist.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library