Skip to Content
Merck

N2644

NωNitro-L-arginine benzyl ester p-toluenesulfonate salt

≥95% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H19N5O4 · C7H8O3S
CAS Number:
Molecular Weight:
481.52
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (TLC)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.N[C@@H](CCCNC(=N)N[N+]([O-])=O)C(=O)OCc2ccccc2

InChI

1S/C13H19N5O4.C7H8O3S/c14-11(7-4-8-16-13(15)17-18(20)21)12(19)22-9-10-5-2-1-3-6-10;1-6-2-4-7(5-3-6)11(8,9)10/h1-3,5-6,11H,4,7-9,14H2,(H3,15,16,17);2-5H,1H3,(H,8,9,10)/t11-;/m0./s1

InChI key

DUIFZPVSNBDNLZ-MERQFXBCSA-N



pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library