Skip to Content
Merck

Skip To

MTCP8010

MTC Bio Reservoir

capacity 10 mL, non-sterile, pack of 300 ea (bulk bag)

Synonym(s):

cryo tube box, cryo tube storage box, reagent dish, reagent holder, MTC Bio P9602-NW

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
300 ea
Please contact Customer Service for Availability
3 770,00 CZK

About This Item

NACRES:
NB.12
UNSPSC Code:
41121522
Manufacturer/tradename:
MTC Bio P8010
Capacity:
10 mL
Sterility:
non-sterile

3 770,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

packaging

pack of 300 ea (bulk bag)

manufacturer/tradename

MTC Bio P8010

capacity

10 mL

General description

Reservoir, 10ml non-sterile, bulk pack, 300/cs

Features and Benefits

  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?

Explore all of our products under MTC Bio Reservoir




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library