Key Documents
MTCC2231W
MTC Bio ScrewSeal Microcentrifuge Tubes
self-standing, clear, capacity 2.0 mL, pkg of 1000 ea (individually wrapped)
Synonym(s):
conical tube, plastic tube
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Pricing and availability is not currently available.
material
blue cap
clear
high-density polyethylene cap
polypropylene
screw top cap
self-standing
sterility
sterile
packaging
pkg of 1000 ea (individually wrapped)
manufacturer/tradename
MTC Bio C2231-W
parameter
22,000 × g speed
capacity
2.0 mL
diam. × L
12.8 mm × 46 mm
General description
Screw-cap microtubes, 2.0mL, with O-ring, , molded graduations, caps attached, sterile, individually wrapped, 500/cs
ScrewSeal screw-cap microtubes are made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leak-proof seal from -90°C to +121°C. Sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000×g.
Features and Benefits
  • Ultra-clear polypropylene
  • Molded or non-graduated options
  • Leak-proof O-ring seal
  • Light-blocking amber available
  • Sterile, RNase, DNase, Pyrogen free
Legal Information
MTC is a trademark of MTC Bio
ScrewSeal is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)