Skip to Content
Merck

Skip To

MSST0062

SILu™Lite APOD, Apolipoprotein D human

recombinant, expressed in HEK 293 cells, MS Protein Standard, ≥95% (SDS-PAGE)

Synonym(s):

Apo-D, ApoD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μg
Please contact Customer Service for Availability
17 900,00 CZK

About This Item

NACRES:
NA.24
UNSPSC Code:
23201100
Form:
lyophilized powder
Assay:
≥95% (SDS-PAGE)
Recombinant:
expressed in HEK 293 cells

17 900,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in HEK 293 cells

Quality Segment

assay

≥95% (SDS-PAGE)

form

lyophilized powder

technique(s)

protein mass spectrometry: suitable

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... APOD(347)

General description

SILu™Lite APOD is a recombinant human protein expressed in human 293 cells. It consists of 189 amino acids (including a C-terminal polyhistidine tag), with a calculated molecular mass of 21.69 kDa. SILu™Lite APOD is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Biochem/physiol Actions

Apolipoprotein D is a member of the Apolipoprotein family. Unlike other lipoproteins, which are mainly produced in the liver, apolipoprotein D is mainly produced in the brain and testes. It was found to be a putative biomarker of androgen receptor function in androgen insensitivity syndrome, the most common cause of disorders of sex development. Apolipoprotein D is associated with neurological disorders and nerve injury, especially related to myelin sheath. It was shown to be elevated in a rat model of stroke, and is elevated in patients with schizophrenia, bipolar disorder, and Alzheimer′s disease.

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Other Notes

QAFHLGKCPNPPVQENFDVNKYLGRWYEIEKIPTTFENGRCIQANYSLMENGKIKVLNQELRADGTVNQIEGEATPVNLTEPAKLEVKFSWFMPSAPYWILATDYENYALVYSCTCIIQLFHVDFAWILARNPNLPPETVDSLKNILTSNNIDVKKMTVTDQVNCPKLSDYKDDDDKGHHHHHHHHGGQ

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSST0056MSST0050MSST0018
description

recombinant, expressed in HEK 293 cells, MS Protein Standard, ≥95% (SDS-PAGE)

description

recombinant, expressed in HEK 293 cells, MS Protein Standard

description

recombinant, expressed in HEK 293 cells, MS Protein Standard

description

recombinant, expressed in HEK 293 cells, MS protein standard

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥98% (SDS-PAGE)

recombinant

expressed in HEK 293 cells

recombinant

expressed in HEK 293 cells

recombinant

expressed in HEK 293 cells

recombinant

expressed in HEK 293 cells

form

lyophilized powder

form

liquid

form

liquid

form

lyophilized powder

Gene Information

human ... APOD(347)

Gene Information

human ... APOB48(338)

Gene Information

human ... AKT1(207)

Gene Information

human ... IL6(3569)

shipped in

ambient

shipped in

dry ice

shipped in

dry ice

shipped in

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters