Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.28
UNSPSC Code:
41116107
Pricing and availability is not currently available.
storage temp.
−20°C
Quality Segment
General description
Alanine metabolism known also as Glucose-Alanine cycle or Cahill cycle is the process in which muscles utilize amino acids for their energetic needs.
- Glutamate and Pyruvate are transformed into Alanine and α-ketoglutarate in the muscle, through the actions of Alanine aminotransferase (AST) and Pyridoxal Phosphate.
- The α-ketoglutarate transforms to Glutamate through the actions of glutamate dehydrogenase with an addition of ammonia. But, in this pathway, the ammonia is excluded from the muscles.
- The Alanine passes through the blood stream and into the liver. In the liver, the Alanine and a-ketoglutarate transform to Pyruvate and Glutamate by the actions of Alanine aminotransferase (AST).
- The Pyruvate transforms to glucose by the Cori cycle (Gluconeogenesis) and returns to the muscle from the blood stream.
Packaging
Components:
Each vial is packaged individually and contains 10 mg of the Alanine pathway metabolite standards.
Each vial is packaged individually and contains 10 mg of the Alanine pathway metabolite standards.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description Supplied by IROA Technologies | description analytical standard | description ≥98% (TLC) |
| Quality Level 200 | Quality Level - | Quality Level 100 | Quality Level 200 |
| storage temp. −20°C | storage temp. −20°C | storage temp. - | storage temp. - |
Kit Components Also Available Separately
Product No.
Description
SDS & Pricing
See All (13)
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1
wgk
WGK 3
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





