Skip to Content
Merck

Skip To

MC1000

SigmaPrep spin column kit with break-away tip

storage temp.:20-25°C, pkg of 1 kit

Synonym(s):

dna extraction, dna purification, spin columns

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NB.22
UNSPSC Code:
40161600


packaging

pkg of 1 kit

storage temp.

20-25°C

General description

SigmaPrep spin column kit with break-away tip enables fast and convenient purification of proteins or protein complexes using affinity media.

Features and Benefits

  • Convenient format for both use and storage
  • Compatible with a variety of microcentrifuge tubes
  • The procedure can be carried out without significant loss of affinity medium
  • Break-away tip or end plug ensures easy storage of the used column or incubation without loss of sample

Legal Information

SigmaPrep is a trademark of Sigma-Aldrich Co. LLC




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library