Skip to Content
Merck

Skip To

MAS009

Albumin Quick Test Strips

Sufficient for 10 tests

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.84
UNSPSC Code:
12352106
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

storage condition

(Keep container tightly closed in a dry and well -ventilated place. Keep away from heat and sources of ignition.)

input

urine
serum
plasma

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases; genitourinary disorders

storage temp.

2-8°C

General description

Albumin, the most abundant plasma protein in humans, fulfills various vital physiological roles. Its functions encompass maintaining colloid osmotic pressure and binding essential substances like long-chain fatty acids, bile acids, bilirubin, haematin, calcium, and magnesium. Moreover, serum albumin serves as a reliable prognostic indicator for various medical conditions, making its detection paramount in healthcare. Lowered serum albumin levels often signal the presence of liver disease, nephritic syndrome, malnutrition, protein-losing enteropathies, and associated morbidity and mortality. Conversely, elevated serum albumin levels are linked to dehydration, indicating the versatility of albumin detection in diagnosing and monitoring health issues. Albumin detection extends beyond serum, as it is a critical parameter in the assessment of urine as well. Albuminuria, the presence of albumin in urine, is a key diagnostic marker for a range of conditions. By identifying albuminuria disease, the early signs of renal disease can be detected, particularly in diabetic patients. In the proposal for Albumin detection kits, these assays are highlighted for their fast, sensitive, and reliable measurement of albumin levels in plasma, serum, and urine. These kits provide a convenient means of assessing health, offering timely intervention, and ultimately contributing to improved patient outcomes and comprehensive healthcare management.

Application

Suitable for the detection of albumin in serum, plasma, urine, etc.
The Albumin Quick Test Strips can be used for determining Albumin levels in metabolomics research

Biochem/physiol Actions

Albumin Quick Test Strips are based on the immobilization of an albumin sensitive chromagen onto a pad. Albumin will react with the chromagen and produce a colored product. The intensity of the product color is directly proportional to the albumin concentration in the sample.

Features and Benefits

  • Rapid and Ultra-Sensitive: Achieve quick results with just 20 µL of the sample, enabling semi-quantitative measurements between 0-0.4 g/dL or 0-60 µM (undiluted) Albumin
  • Quick Results: Complete sample treatment and the assay in under 15 minutes, saving you valuable time
  • User-Friendly Design: Our convenient test strip design eliminates the requirement for a plate or cuvette reader, making the testing process simple and hassle-free

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9056A8577A7034
Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

detection method

colorimetric

detection method

-

detection method

-

detection method

-

input

urine
serum
plasma

input

-

input

-

input

-

storage condition

(Keep container tightly closed in a dry and well -ventilated place. Keep away from heat and sources of ignition.)

storage condition

-

storage condition

-

storage condition

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

relevant disease(s)

gastrointestinal diseases; genitourinary disorders

relevant disease(s)

-

relevant disease(s)

-

relevant disease(s)

-


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

55.4 °F

flash_point_c

13 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters