Skip to Content
Merck

Skip To

MAK511

Malate Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Malate Quantification Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
12 200,00 CZK

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503

12 200,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

application(s)

agriculture
food and beverages

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

General description

L-malic acid, or L-malate, is a dicarboxylic acid that is made by all living organisms and plays an important role in the Calvin and Krebs Cycles. It is a source of CO2 for the Calvin cycle in plants and is also an intermediate that forms from fumarate in the Krebs Cycle. Malate is frequently used in food and beverage industries as an additive in products such as wine, beer, candies, etc.

Application

The kit is suitable for the determination of malate in food, juice, beverage and other agricultural products.

Biochem/physiol Actions

The Malate Assay Kit is based on the malate dehydrogenase catalyzed oxidation of malate in which the formed NADH reduces a formazan (MTT) reagent. The intensity of the product color, measured at 565 nm is proportional to the malate concentration in the sample.

Features and Benefits

  • Linear detection range 0.02 to 2 mM
  • Compatible with high-throughput handling systems.



Still not finding the right product?

Explore all of our products under Malate Assay Kit


Storage Class

10 - Combustible liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library