MAK398
5′-Nucleotidase (CD73) Activity Assay Kit (Colorimetric)
sufficient for 100 colorimetric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
detection method
colorimetric
relevant disease(s)
gastrointestinal diseases
storage temp.
−20°C
General description
5′-Nucleotidase (5′-NT), also known at CD73 is an enzyme located in the plasma membrane. It converts extracellular nucleotides like 5′-AMP to their corresponding nucleosides, through phosphorylitic cleavage. This conversion facilitates uptake of the nucleosides through nucleoside receptors into the cell, where they can again be phosphorylated to generate nucleotides and contribute to the nucleotide pool, inside the cell. 5′-NT levels are elevated in hepatic diseases such as viral hepatitis, alcoholic liver disease and cirrhosis.
Features and Benefits
5′-Nucleotidase (CD73) Activity Assay Kit:
- Sensitive, detecting as low as 0.2 mU of 5′-NT.
- Compatible with high-throughput handling systems.
Suitability
The kit is suitable for the measurement of 5′-Nucleotidase activity in animal tissue lysates (eg. liver), cell lysates, recombinant enzyme and purified protein.
Principle
The 5′-Nucleotidase (CD73) Activity Assay Kit is a simple two-step end point assay that relies on the Berthelot′s test for quantification of ammonia. In this assay, the action of 5′-nucleotidase on the substrate generates a product, which releases ammonia in presence of the converter. Developer I and II are then used to quantify the released ammonia through increase in absorbance at A670 nm. This assay can detect as low as 0.2 mU of 5′-NT. Since non-specific enzymes like alkaline phosphatase can give a positive signal in this assay, 5′-NT inhibitor may be used to completely inhibit 5′-nucleotidase and distinguish from the signal from non-specific enzymes. The assay kit also includes 5′- Nucleotidase (5′-NT) enzyme for use as a positive control.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 1A Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation
Target Organs
Lungs
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service