Skip to Content
Merck
All Photos(1)

Documents

MAK276

Sigma-Aldrich

Sulfatase Activity Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Sulfatases (EC 3.1.6) are a family of enzymes that catalyze the hydrolysis of sulfate ester bonds from a broad range of biological molecules, including steroids, carbohydrates, and proteins. They can be found in intracellular and extracellular spaces and are distributed in a wide range of cells and tissues. Intracellular sulfatases are commonly found localized within the lysosome. Genetic defects in sulfatase can result in certain lysosomal storage disorders and abnormal expression can contribute to certain hormone-dependent cancers, such as breast and prostate cancer.

Suitability

Suitable for measuring activity of purified enzyme as well as sulfatase from biological samples.

Principle

This Sulfatase Activity Assay Kit provides a quick and easy way to determine sulfatase activity. The kit measures the hydrolysis of a sulfate ester to 4-nitrocatechol, which can be detected at 515 nm. The kit is suitable for measuring activity of purified enzyme as well as sulfatase from biological samples. The limit of detection is below 1 mU.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service