MAK272
High Sensitivity β-Hydroxybutyrate Fluorometric Assay Kit
sufficient for 100 fluorometric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.25
Recommended Products
application(s)
cosmetics
food and beverages
detection method
fluorometric
relevant disease(s)
endocrinological disorders, diabetes; gastrointestinal diseases
storage temp.
−20°C
General description
β -Hydroxybutyrate (β-HB), also known as 3-hydroxybutyrate, is a ketone body primarily synthesized in the liver from the oxidation of fatty acids. Ketone body formation, ketogenesis, is elevated when blood glucose levels drop. Moderately elevated blood ketone bodies occur normally during fasting and prolonged exercise. During alcoholic or diabetic ketoacidosis, blood levels of β -Hydroxybutyrate can rise to abnormally high levels. β -Hydroxybutyrate can be used as a marker for hepatic energy metabolism.
Suitability
Suitable for use with serum, plasma, urine, and other biological fluids, as well as food products such as milk.
Principle
The High Sensitivity β-Hydroxybutyrate Assay Kit is a simple, sensitive, high-throughput adaptable method for quantifying β-hydroxybutyrate when sample quantities are limited or sub-ketogenic levels are suspected. In this assay, β-hydroxybutyrate is enzymatically oxidized generating a fluorometric signal (lex = 535 nm/lem = 587 nm), proportional to the amount of β-hydroxybutyrate present. The assay is sensitive to 4 mM of β-hydroxybutyrate in a variety of biological samples.
replaced by
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service