Skip to Content
Merck

Skip To

MAK261

Glycine Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Synonym(s):

Fluorometric Glycine Detection Kit

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503


detection method

fluorometric

relevant disease(s)

cancer; neurological disorders

storage temp.

−20°C

General description

Glycine (GLY) is a non-essential amino acid used to help create muscle tissue and convert glucose into energy. It′s basic functions include the participation in the synthesis of creatine, glutathione, heme groups, and conjugated bile acids (bile salts). It is also used in the synthesis of DNA and RNA. It is one of the most abundant residues in the triple-helical structure of collagen, which contributes to the elasticity of skin and connective tissue. It acts as a glucogenic amino acid regulating blood sugar levels. Glycine is necessary for central nervous system function. It possesses both inhibitory and excitatory neurotransmitter functions in the brain stem and spinal cord. Recent studies suggest, glycine may play a role in tumorigenesis and malignancy. Some of the external sources of glycine includes legumes, meat, fish and dairy products.

Application

Suitable for the detection of glycine in biological fluids such as serum, plasma, saliva, urine, etc

Biochem/physiol Actions

The Glycine Assay Kit is a simple and reliable, high throughput assay that detects the physiological concentration of glycine in a variety of biological fluids. In the assay, glycine is oxidized generating a fluorometric (lex = 535 nm/ lem = 587 nm) product, proportional to the glycine present. The assay is specific, and other standard and non-standard amino acids do not interfere with the assay. The kit is sensitive to 1 mM of glycine.

Features and Benefits

Compatible with high-throughput handling systems.


signalword

Danger

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3

Hazard Classifications

Aquatic Chronic 2 - ED ENV 1 - Met. Corr. 1 - Resp. Sens. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters