Skip to Content
Merck

Skip To

M9307

Anti-Macrophage-Derived Chemokine antibody produced in chicken

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-MDC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.1 mg
Please contact Customer Service for Availability
CZK 17,200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.21
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

CZK 17,200.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL22(6367)

Immunogen

recombinant human MDC expressed in E. coli.

Biochem/physiol Actions

This antibody will neutralize the biological activity of recombinant human MDC. The antibody shows 20% cross-reactivity with recombinant human MPIF-1, 10% cross-reactivity with recombinant human MIP-1δ and 1% cross-reactivity with recombinant mouse MDC, human HCC-1, human I-309, mouse MIP-1α, viral MIP-I, mouse MIP-1β and mouse MIP-1γ.

Physical form

Lyophilized from a 0.2 μm-filtered solution in phosphate buffered saline with 5% Trehalose.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1304901M2420
Quality Level

200

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

purified immunoglobulin

biological source

chicken

biological source

-

biological source

mouse

clone

polyclonal

clone

polyclonal

clone

24822, monoclonal

UniProt accession no.

O00626

UniProt accession no.

O00626

UniProt accession no.

P13500


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters