Skip to Content
Merck
All Photos(1)

Key Documents

M8031

Sigma-Aldrich

Methyl 2,3,4,6-tetra-O-acetyl-β-D-thiogalactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O9S
CAS Number:
Molecular Weight:
378.39
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CS[C@@H]1O[C@H](COC(C)=O)[C@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O

InChI

1S/C15H22O9S/c1-7(16)20-6-11-12(21-8(2)17)13(22-9(3)18)14(23-10(4)19)15(24-11)25-5/h11-15H,6H2,1-5H3/t11-,12+,13+,14-,15+/m1/s1

InChI key

XWFUCHLBRWBKGN-FQKPHLNHSA-N

Application

Methyl 2,3,4,6-tetra-O-acetyl-β-D-thiogalactopyranoside has been used in a study to assess applications of ionic liquids in carbohydrate chemistry. It has also been used to study thioglycosylation in ionic liquids.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianguo Zhang et al.
Beilstein journal of organic chemistry, 2, 12-12 (2006-06-29)
A novel, green chemistry, glycosylation strategy was developed based upon the use of ionic liquids. Research studies demonstrated that thiomethyl glycosides could readily be activated with methyl trifluoromethane sulfonate, using 1-butyl-3-methylimidazolium tetrafluoroborate as a solvent. This green chemistry glycosylation strategy
Omar A El Seoud et al.
Biomacromolecules, 8(9), 2629-2647 (2007-08-19)
Ionic liquids (ILs) are composed only of ions. Of special interest to this review are those where at least one ion (the cation) is organic and whose melting points are below or not far above room temperature. ILs are designated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service