Skip to Content
Merck

M5168

Magnesium perchlorate

ACS reagent, passes test for moisture absorption

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Mg(ClO4)2
CAS Number:
Molecular Weight:
223.21
EC Number:
233-108-3
UNSPSC Code:
12352300
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

impurities

≤0.005 meq/g Titratable free acid, ≤0.025 meq/g Titratable base

loss

≤8% loss on drying, 190°C

suitability

passes test for moisture absorption

SMILES string

[Mg++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Mg/c2*2-1(3,4)5;/h2*(H,2,3,4,5);/q;;+2/p-2

InChI key

MPCRDALPQLDDFX-UHFFFAOYSA-L



Still not finding the right product?

Explore all of our products under Magnesium perchlorate


pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library