Skip to Content
Merck

M3789

Methyl 12-methylmyristate

Synonym(s):

12-Methyltetradecanoic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H32O2
CAS Number:
Molecular Weight:
256.42
UNSPSC Code:
12352211
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>97% (GC)

form

liquid

functional group

ester

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCC(C)CCCCCCCCCCC(=O)OC

InChI

1S/C16H32O2/c1-4-15(2)13-11-9-7-5-6-8-10-12-14-16(17)18-3/h15H,4-14H2,1-3H3

InChI key

BJIUDNXPLSJWKE-UHFFFAOYSA-N



Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Guy Schleyer et al.
Nature microbiology, 4(3), 527-538 (2019-02-06)
Tapping into the metabolic crosstalk between a host and its virus can reveal unique strategies employed during infection. Viral infection is a dynamic process that generates an evolving metabolic landscape. Gaining a continuous view into the infection process is highly
Simone Hauff et al.
Journal of agricultural and food chemistry, 58(2), 707-712 (2010-01-02)
Branched chain fatty acids (iso- and anteiso-fatty acids) are common minor compounds of the lipids found in dairy products and fish (<1-3%) and major fatty acids of Gram-positive bacteria. Their presence in food has been associated with bacterial sources. Bacterial
Simone Hauff et al.
Lipids, 45(4), 357-365 (2010-03-12)
Anteiso fatty acids (aFA) are substituted with a methyl group on the antepenultimate carbon of the straight acyl chain. This feature leads to a stereogenic center. The 12-methyltetradecanoic acid (a15:0) and the 14-methylhexadecanoic acid (a17:0) are the most common aFA