Skip to Content
Merck

M2641

Methyl-[5-(N-methyl-N-phenylamino)-2,4-pentadienylidene]phenylammonium chloride

Dye content ~90 %

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C19H21N2Cl
CAS Number:
Molecular Weight:
312.84
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
237-774-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


composition

Dye content, ~90%

mp

117-121 °C (lit.)

SMILES string

[Cl-].CN(\C=C\C=C\C=[N+](/C)c1ccccc1)c2ccccc2

InChI

1S/C19H21N2.ClH/c1-20(18-12-6-3-7-13-18)16-10-5-11-17-21(2)19-14-8-4-9-15-19;/h3-17H,1-2H3;1H/q+1;/p-1

InChI key

FPYATAHHYUQBFA-UHFFFAOYSA-M



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library