Skip to Content
Merck

Skip To

M2199

PMEG hydrate

≥98% (HPLC)

Synonym(s):

9-[2-(Phosphonomethoxy)ethyl]guanine hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H12N5O5P · xH2O
CAS Number:
Molecular Weight:
289.19 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

O.NC1=Nc2c(ncn2CCOCP(O)(O)=O)C(=O)N1

InChI

1S/C8H12N5O5P.H2O/c9-8-11-6-5(7(14)12-8)10-3-13(6)1-2-18-4-19(15,16)17;/h3H,1-2,4H2,(H2,15,16,17)(H3,9,11,12,14);1H2

InChI key

BDVQXHFCXJXVND-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PZ0158D6196SML2774
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: ≥5 mg/mL

solubility

DMSO: ≥45 mg/mL

solubility

H2O: ≥10 mg/mL

solubility

H2O: 2 mg/mL, clear

storage condition

desiccated

storage condition

desiccated

storage condition

-

storage condition

desiccated

Biochem/physiol Actions

The acyclic nucleotide 9-(2-phosphonylmethoxyethyl)guanine (PMEG) forms an active phosphorylated metabolite, PMEG diphosphate (PMEGpp), in cells and causes cytotoxicity in dividing cells due to potent inhibition of the nuclear DNA polymerases resulting in inhibition of DNA synthesis and/or DNA repair.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernhard Ellinger et al.
Viruses, 12(5) (2020-05-30)
HIV-1 infection is a complex, multi-step process involving not only viral, but also multiple cellular factors. To date, drug discovery methods have primarily focused on the inhibition of single viral proteins. We present an efficient and unbiased approach, compatible with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service