Skip to Content
Merck

M200

(±)-N-Methyl-1-(3,4-methylenedioxyphenyl)-2-butanamine hydrochloride

solid

Synonym(s):

(±)-N-Methyl-1,3-benzodioxole-5-butanamine hydrochloride, MBDB hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C12H17NO2 · HCl
CAS Number:
Molecular Weight:
243.73
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

solid

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada, kontrollierte Droge in Deutschland

color

white

solubility

H2O: soluble

SMILES string

Cl.CCC(Cc1ccc2OCOc2c1)NC

InChI

1S/C12H17NO2.ClH/c1-3-10(13-2)6-9-4-5-11-12(7-9)15-8-14-11;/h4-5,7,10,13H,3,6,8H2,1-2H3;1H

InChI key

LFXXIXAVEJVYGY-UHFFFAOYSA-N

Biochem/physiol Actions

Non-hallucinogenic; CNS stimulant. Analog of MDMA

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryam Tabasinezhad et al.
Protein expression and purification, 155, 59-65 (2018-11-24)
Recombinant antibodies have emerged over the last few decades as the fastest growing class of therapeutic proteins for autoimmune diseases. Post-translation modifications of antibodies produced by human cell lines are highly consistent with those existing in natural human proteins and
M P Johnson et al.
European journal of pharmacology, 200(1), 9-16 (1991-07-23)
The ability of several 3,4-methylenedioxymethamphetamine (MDMA) analogues to inhibit the uptake of [3H]serotonin (5-HT), dopamine (DA) and norepinephrine (NE) into synaptosomes was examined. In addition, the ability of the compounds to inhibit the uptake of [3H]5-HT and DA into synaptosomes
Fengxia Lin et al.
Drug design, development and therapy, 14, 621-633 (2020-02-28)
Dysfunction of endothelial cells plays a key role in the pathogenesis of diabetic atherosclerosis. High glucose (HG) has been found as a key factor in the progression of diabetic complications, including atherosclerosis. PI3K/Akt/eNOS signaling pathway has been shown to involve
S C Dulawa et al.
Psychopharmacology, 149(3), 306-312 (2000-05-24)
Prepulse inhibition (PPI) is the normal reduction of the startle response which occurs when an abrupt startling stimulus is preceded by a weak pre-stimulus and is decreased in several neuropsychiatric disorders. The role of the serotonin 1B (5-HT(1B)) receptor in
Ya-Ying Chang et al.
Inflammopharmacology, 27(2), 249-260 (2019-02-06)
We investigated effects of magnesium sulfate (MgSO4) on modulating lipopolysaccharide (LPS)-macrophage binding and cluster of differentiation 14 (CD14) expression. Flow cytometry data revealed that the mean levels of LPS-macrophage binding and membrane-bound CD14 expression (mCD14) in differentiated THP-1 cells (a

Related Content

Global Trade Item Number

SKUGTIN
775266-100MG04061833231531

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service