Skip to Content
Merck

M1938

8-Methoxypyrene-1,3,6-trisulfonic acid trisodium salt

Synonym(s):

PTS1, Trisodium 8-methoxypyrene-1,3,6-trisulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H9Na3O10S3
CAS Number:
Molecular Weight:
538.41
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

253-260 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[Na+].COc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C17H12O10S3.3Na/c1-27-12-6-13(28(18,19)20)9-4-5-11-15(30(24,25)26)7-14(29(21,22)23)10-3-2-8(12)16(9)17(10)11;;;/h2-7H,1H3,(H,18,19,20)(H,21,22,23)(H,24,25,26);;;/q;3*+1/p-3

InChI key

GUGNSJAORJLKGP-UHFFFAOYSA-K





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library