M0814
R(−)-Me5 hydriodide
solid
Synonym(s):
1-(2,6-Dimethylphenoxy)-3-methyl-2-butanamine hydriodide
About This Item
form
solid
color
white
solubility
H2O: 22 mg/mL
SMILES string
I[H].CC(C)C(N)COc1c(C)cccc1C
InChI
1S/C13H21NO.HI/c1-9(2)12(14)8-15-13-10(3)6-5-7-11(13)4;/h5-7,9,12H,8,14H2,1-4H3;1H
InChI key
AQLAXCGHPBMFIJ-UHFFFAOYSA-N
Gene Information
human ... SCN2A(6326)
Application
Biochem/physiol Actions
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service