Skip to Content
Merck

M0185

Anti-MyD88, Internal Domain antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35 kDa

species reactivity

human, mouse

concentration

~0.5 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using human Jurkat cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MYD88(4615)
mouse ... Myd88(17874)

Immunogen

synthetic peptide corresponding to amino acids 233-248 of the internal domain of human MyD88. This peptide differs from mouse MyD88 by 2 amino acids.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


T P Bonnert et al.
FEBS letters, 402(1), 81-84 (1997-01-27)
Murine MyD88, an RNA with homology both to the interleukin-1 receptor signaling domain and to 'death-domains', is rapidly upregulated during differentiation of the myeloleukemic cell line M1. We have cloned the human homologue of murine MyD88 and re-evaluated the murine