Skip to Content
Merck

L9384

L-Leucine 3-carboxy-4-hydroxyanilide hydrochloride

suitable for ligand binding assays

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H18N2O4 · HCl
CAS Number:
Molecular Weight:
302.75
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Leucine 3-carboxy-4-hydroxyanilide hydrochloride,

technique(s)

ligand binding assay: suitable

storage temp.

2-8°C

SMILES string

Cl.CC(C)C[C@H](N)C(=O)Nc1ccc(O)c(c1)C(O)=O

InChI

1S/C13H18N2O4.ClH/c1-7(2)5-10(14)12(17)15-8-3-4-11(16)9(6-8)13(18)19;/h3-4,6-7,10,16H,5,14H2,1-2H3,(H,15,17)(H,18,19);1H/t10-;/m0./s1

InChI key

BSVJWFHAORBGDY-PPHPATTJSA-N

Application

L-Leucine 3-carboxy-4-hydroxyanilide may be used as a substrate for determining the activity of microsomal aminopeptidase in serum.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library