Skip to Content
Merck
All Photos(1)

Key Documents

L5390

Sigma-Aldrich

Lectin from Erythrina cristagalli (coral tree)

lyophilized powder

Synonym(s):

Erythrina cristagalli agglutinin, ECA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 7,730.00

CZK 7,730.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
CZK 7,730.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

CZK 7,730.00


Please contact Customer Service for Availability

form

lyophilized powder

Quality Level

potency

<0.5 μg per mL agglutination activity

composition

Protein, ≥85% biuret

impurities

salt, free

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

ECA is not blood group specific, but has an affinity for D-galactose and D-galactosides. It is reported to be mitogenic for human peripheral blood T lymphocytes.
Lectin or haemagglutinin[1] is a sugar stereospecific binding protein or glycoprotein. It is capable of reversibly agglutinating cells. It is implicated in various cellular processes such as cell agglutination, homing of leukocytes, induction of mitosis in lymphocytes, interferon c (IFN c) and cytotoxicity.[2] It is used as a blood typing reagent in immunohaematology. Lectin is also used as a reagent in testing erythrocyte polyagglutinable states. It is considered as a potential tool for analyzing cell surface architecture and related membrane functions.[1]

Preparation Note

Purified by affinity chromatography.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of trypsinized human blood group O erythrocytes after 1 hr at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human erythrocyte specific lectin from the seeds of Indian coral tree, Erythrina variegata Linn, var. orientali Linn, Merrill
Datta TK and Basu PS
Journal of Biosciences, 5(1), 25-30 (1983)
Purification, some properties of a D-galactose-binding leaf lectin from Erythrina indica and further characterization of seed lectin
Konozy EHE, et al.
Biochimie, 84(10), 1035-1043 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service