Skip to Content
Merck
All Photos(1)

Documents

K4018

Sigma-Aldrich

PKCζ, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

97-131 nmol/min·mg

mol wt

~93 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCZ(5590)

Biochem/physiol Actions

PKCζ (protein kinase C ζ) is a atypical PKC and works independent of Ca2+ and diacylglycerol. It is involved in MAPK (mitogen-activated protein kinase)-induced nuclear factor κ B activation. It also stabilizes DNA mismatch-repair proteins, participates in laeukemic cell differentiation, cell migration and chemotaxis. PKCζ have tumour suppressor as well as oncogenic properties. Upregulation of PKCζ causes apoptosis in ovarian carcinoma cells. On the other hand, absence of PKCζ decreases tumorigenic activity in a colon cancer mouse model.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Kim et al.
Biochemical and biophysical research communications, 237(2), 336-339 (1997-08-18)
To examine whether multiple protein kinase C (PKC) isoforms could interact or crosstalk, phorbol ester-insensitive atypical PKC (aPKC) zeta isoform was overexpressed in NIH3T3 cells, and the cells were stimulated with phorbol ester to activate diacylglycerol-dependent conventional (cPKC) and novel
A Romanelli et al.
Molecular and cellular biology, 19(4), 2921-2928 (1999-03-19)
p70 S6 kinase (p70S6K) is an important regulator of cell proliferation. Its activation by growth factor requires phosphorylation by various inputs on multiple sites. Data accumulated thus far support a model whereby p70S6K activation requires sequential phosphorylations at proline-directed residues
S A Hartsink-Segers et al.
Leukemia, 29(2), 304-311 (2014-07-06)
Both tumour suppressor and oncogenic functions have been ascribed to the atypical zeta isoform of protein kinase C (PKCζ), whereas its constitutively active form PKMζ is almost exclusively expressed in the brain where it has a role in long-term memory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service