K2893
PAK1/CDC42, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
MGC130000, MGC130001, PAKalpha
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51111800
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
40-53 nmol/min·mg
mol wt
~96 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PAK1(5058)
Biochem/physiol Actions
PAK1 is a member of the p21-activated kinases (PAKs) which have been implicated in the regulation of gene transcription, cell morphology, motility and apoptosis. PAKs are serine/threonine kinases which bind to and are activated by small GTPases, CDC42 (cell division cycle 42) and RAC. PAK1 belongs to the Group I PAKs which also includes PAK2 and PAK3. PAK1 is a key regulator of the actin cytoskeleton, adhesion and cell motility.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M C Parrini et al.
Biochemical Society transactions, 33(Pt 4), 646-648 (2005-07-27)
Pak1 (p21-activated kinase 1) is a key regulator of the actin cytoskeleton, adhesion and cell motility. Such biological roles require a tight spatial and kinetic control of its localization and activity. We summarize here the current knowledge on Pak1 dynamics
Zahara M Jaffer et al.
The international journal of biochemistry & cell biology, 34(7), 713-717 (2002-04-16)
The p21-activated kinases (Paks) are serine/threonine protein kinases that bind to and, in some cases, are stimulated by activated forms of the small GTPases, Cdc42 and Rac. With the recent discovery of several novel isoforms, Paks are now categorized into
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service