Skip to Content
Merck

K1519

KM11060

≥98% (HPLC)

Synonym(s):

7-Chloro-4-[4-[4-chlorophenyl)sulfonyl]-1-piperazinyl]quinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H17Cl2N3O2S
CAS Number:
Molecular Weight:
422.33
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

Clc1ccc(cc1)S(=O)(=O)N2CCN(CC2)c3ccnc4cc(Cl)ccc34

InChI

1S/C19H17Cl2N3O2S/c20-14-1-4-16(5-2-14)27(25,26)24-11-9-23(10-12-24)19-7-8-22-18-13-15(21)3-6-17(18)19/h1-8,13H,9-12H2

InChI key

GIEHIZKCIZLXLF-UHFFFAOYSA-N

Biochem/physiol Actions

KM11060 is a mutated F508del CFTR corrector.
KM11060 is a mutated F508del cystic fibrosis transmembrane conductance regulator (CFTR) corrector. The most common mutation (~90%) in the CFTR gene is a deletion of phenylalanine 508 (F508del). The mutation impairs folding, trafficking, membrane stability, and channel gating leading to reduced CFTR expression and chloride conductance in the apical membrane and other abnormalities. KM11060 is an analog of sildenafil, which restores a function of the F508del mutated CFTR chloride channel. KM11060 appears to be more potent than sildenafil, forskolin, and genistein.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library