Skip to Content
Merck
All Photos(1)

Key Documents

K0879

Sigma-Aldrich

Kanamycin sulfate Streptomyces kanamyceticus

powder, γ-irradiated

Synonym(s):

Kanamycin sulfate from Streptomyces kanamyceticus, Kanamycin A, Kanamycin sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.85

biological source

Streptomyces kanamyceticus

grade

Molecular Biology
for molecular biology

sterility

γ-irradiated

form

powder

potency

≥700 μg per mg

color

white to off-white

pH

6.5-8.5(1% solution)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

suitability

nonselective for Escherichia coli
nonselective for coliforms

SMILES string

OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1

InChI key

OOYGSFOGFJDDHP-KMCOLRRFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside
Kanamycin, A from Streptomyces kanamyceticus[1] is a polycationic aminoglycoside[1] and a broad-spectrum antibiotic.[2] Structurally, it comprises hydroxyl and amino functional groups.[1]

Application

Kanamycin sulfate Streptomyces kanamyceticus has been used as an antibiotic component in Murashige and Skoog medium for the selection of tobacco tissue culture explants.[3] It has also been used as an antibiotic in Luria broth for culturing Escherichia coli K-12 strain MG1655 cells for secretion assay[4] and E. coli TV1061 cells for immobilization studies.[5]

Biochem/physiol Actions

Kanamycin′s hydroxyl and amino functional groups are crucial for its membrane action functionality. However, it also exhibits toxicity towards mammalian cells.[1] It prevents contamination in cell culture as well as a selective agent for bacteria with kanamycin resistance.
Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance:Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Principle

Kanamycin binds to the 30S ribosomal subunit and inhibits protein synthesis, leading to cell death.

Preparation Note

Prepare stock solution directly in the vial with sterile water to a final concentration of 10mg/mL. Stock solution should be stored at 2-8°C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin M Krause et al.
Cold Spring Harbor perspectives in medicine, 6(6) (2016-06-03)
Aminoglycosides are natural or semisynthetic antibiotics derived from actinomycetes. They were among the first antibiotics to be introduced for routine clinical use and several examples have been approved for use in humans. They found widespread use as first-line agents in
Shoot Regenerative Capacity Assays in Arabidopsis and Tobacco
Tian-QiZ and Jia-Wei W
Bio-protocol, 6(5) (2016)
Torsten John et al.
Biochimica et biophysica acta. Biomembranes, 1859(11), 2242-2252 (2017-08-30)
Biological membranes are natural barriers to the transport of molecules and drugs within human bodies. Many antibacterial agents need to cross these membranes to reach their target and elicit specific effects. Kanamycin A belongs to the family of aminoglycoside antibiotics
Tim Axelrod et al.
Talanta, 149, 290-297 (2016-01-01)
Toxicants in water sources are of concern. We developed a tool that is affordable and easy-to-use for monitoring toxicity in water. It is a biosensor composed of disposable bioreporter pads (calcium alginate matrix with immobilized bacteria) and a non-disposable CMOS
Atef Allam et al.
Journal of immunology (Baltimore, Md. : 1950), 193(2), 871-878 (2014-06-11)
The role of the TNF family member CD70 in adaptive T cell responses has been intensively studied, but its function in innate responses is still under investigation. In this study, we show that CD70 inhibits the early innate response to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service