Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C8H15NO4 · C7H8SO3
CAS Number:
Molecular Weight:
361.41
UNSPSC Code:
12352201
MDL number:
storage temp.
−20°C
SMILES string
Cc1ccc(cc1)S(O)(=O)=O.CC2(C)O[C@H]3[C@H](N)O[C@H](CO)[C@H]3O2
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.