Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3CH2CH(CH3)CH(NH2)COOH
CAS Number:
Molecular Weight:
131.17
EC Number:
207-139-8
UNSPSC Code:
12352207
PubChem Substance ID:
Beilstein/REAXYS Number:
1721790
MDL number:
technique(s)
cell culture | mammalian: suitable
impurities
leucine, essentially free
mp
290 °C (dec.) (lit.)
SMILES string
CC[C@@H](C)[C@@H](N)C(O)=O
InChI
1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m1/s1
InChI key
AGPKZVBTJJNPAG-RFZPGFLSSA-N
General description
This product is a mixture of four stereoisomers. Isoleucine is an essential amino acid. Isoleucine is a branched-chain amino acid (BCAA) that is frequently used in combination with the other branched chain amino acids, valine and leucine.
Other Notes
A mixture of 4 stereoisomers.
Still not finding the right product?
Explore all of our products under
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.