Skip to Content
Merck

Skip To

I5396

IL-1α mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Interleukin-1α, mIL-1α, IL-1α

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 μg
Please contact Customer Service for Availability
CZK 11,200.00
25 μg
Please contact Customer Service for Availability
CZK 30,300.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Biological source:
mouse
Recombinant:
expressed in E. coli
Assay:
≥97% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
18 kDa
Impurities:
<0.1 EU/μg

CZK 11,200.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Interleukin-1α from mouse, IL-1α, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

mouse

Quality Segment

recombinant

expressed in E. coli

assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

3-7 pg/mL ED50

quality

endotoxin tested

mol wt

18 kDa

packaging

pkg of 5 and 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... IL1A(3552)

Biochem/physiol Actions

Interleukin-1 (IL-1), originally known as Lymphocyte Activating Factor (LA F), activates T cells and lymphocytes, which then proliferate and secrete Interleukin-2 (IL-2). The proliferative activity of human IL-1α is tested in culture using the murine cell line D10S.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.25 mg bovine serum albumin.

Analysis Note

The biological activity is measured in a cel proliferation assay using the murine helper T cell line, D10.G4.1.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I4026I9646I5271
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

assay

≥97% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?

Explore all of our products under Interleukin-1α from mouse


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters