Skip to Content
Merck

I3880

4-(Iodoacetamido)salicylic acid

~98%, crystalline

Synonym(s):

4-(Iodoacetamido)-2-hydroxybenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H8INO4
CAS Number:
Molecular Weight:
321.07
UNSPSC Code:
12352103
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~98%

form

crystalline

color

tan to brown

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(NC(=O)CI)cc1O





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library