Skip to Content
Merck

Skip To

I3400

Interferon-γ-Inducible Protein-10 human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

CXCL10, IP-10

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg
Please contact Customer Service for Availability
CZK 5,910.00
CZK 5,023.50

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32
MDL number:

CZK 5,023.50

List PriceCZK 5,910.00Save 15%
Web-Only Promotion

Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.015-0.075 μg/mL ED50 (chemoattract mouse BaF3 cells transfected with hCXCR3), 0.03-0.18 μg/mL ED50 (chemoattract T-Lymphocytes)

mol wt

8.7 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CXCL10(3627)

General description

The gene for C-X-C motif chemokine ligand 10 (CXCL10), also known as interferon-γ-inducible protein-10 (IP-10), is mapped to human chromosome 4q21.1.[1] IP-10 is secreted by monocytes, neutrophils, endothelial cells, keratinocytes, fibroblasts, mesenchymal cells, dendritic cells, astrocytes and hepatocytes.[2] It is a polypeptide of 78 amino acids. Human IP-10 shows 67% amino acid homology to mouse CRG-2.

Biochem/physiol Actions

IP-10 (interferon-γ-inducible protein-10) protein is a proinflammatory chemokine which controls immune responses. It interacts with chemokine receptor 3 (CXCR3) and activates leukocytes including T cells, eosinophils, monocytes and natural killer cells. Mutations in IP-10 are associated with rheumatoid arthritis.[3] In humans with chronic hepatitis C virus (HCV) infection or HCV/human immunodeficiency virus (HIV) co-infection, IP-10 is linked with obesity and insulin resistance.[2]
IP-10, a chemoattractant for activated T lymphocytes, is a potent inhibitor of angiogenesis and displays thymus-dependent anti-tumor effects.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured by its ability to chemoattract mouse BaF/3 cells transfected with hCXCR3.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I5528SRP3246H7541
assay

≥97% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

assay

≥98% (HPLC), ≥98% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

biological source

human

biological source

human

biological source

rat

biological source

human

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in HEK 293 cells

Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

100

form

lyophilized powder

form

lyophilized powder

form

lyophilized

form

lyophilized powder

packaging

pkg of 10 μg

packaging

pkg of 25 μg

packaging

pkg of 25 μg

packaging

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters