Skip to Content
Merck
All Photos(1)

Key Documents

I2533

Sigma-Aldrich

Anti-phospho-Insulin Receptor Substrate-1 (pSer312) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-phospho-IRS-1(pSer312)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

western blot: 1:1000 using stimulated CHO cell line

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer312)

Gene Information

human ... IRS1(3667)
mouse ... Irs1(16367)
rat ... Irs1(25467)

General description

Insulin receptor substrate-1 (IRS-1) is among the four IRS proteins (IRS-1-IRS-4) that mediate signalling by insulin receptor (IR) and insulin-like growth factor (IGF) receptor. IRS-1 has a pleckstrin homology domain and a phosphotyrosine-binding domain. Phosphorylation of the Tyr1179 residue in the SH-1 negatively regulates insulin. IRS-1 has the abililty to translocate to the nucleus, the mechanism of which is not very clear. Estrogen receptors (ER) α and β have a strong affinity to IRS-1, an important interaction in breast cancers and medulloblastomas. The expression of IRS-1 in prostate cancer is associated with lower tumor cell invasiveness. Along with ER α, IRS-1 also modulates transcriptional activity involved in the development of breast and prostate cancer. The signalling pathway involving IGF-IR-IRS-1 in cells expressing JCV T-antigen has been reported to affect the proliferation, survival and DNA repair in the cells. Phosphorylation of IRS-1 at Ser312 and Ser616 results in the negative regulation of insulin signalling in humans
Anti-phospho-IRS-1 (pSer312) specifically recognizes human IRS-1 (pSer312). It may react with mouse and rat (100% homology) IRS-1.

Immunogen

synthetic phosphopeptide derived from the region of IRS-1 that contains serine 312. This sequence is 100% homologous with mouse and rat IRS-1.

Application

Anti-phospho-IRS-1 (pSer312) antibody may be used for detection by immunoblotting, in stimulated CHO cell line expressing human insulin receptor, at a working dilution of 1:1000.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 containing 50% glycerol, 1.0 mg/mL BSA (IgG, protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Aguirre et al.
The Journal of biological chemistry, 277(2), 1531-1537 (2001-10-19)
Serine phosphorylation of insulin receptor substrate-1 (IRS-1) inhibits insulin signal transduction in a variety of cell backgrounds, which might contribute to peripheral insulin resistance. However, because of the large number of potential phosphorylation sites, the mechanism of inhibition has been
Joanna Trojanek et al.
Journal of cellular physiology, 206(1), 35-46 (2005-06-21)
JC polyomavirus (JCV), which infects 90% of the human population, is detectable in human tumors. Its early protein, JCV T-antigen, transforms cells in vitro and is tumorigenic in experimental animals. Although T-antigen-mediated transformation involves genetic alterations of the affected cells
Sindhu Saraswathy et al.
Experimental eye research, 196, 108064-108064 (2020-05-23)
This work sought to compare aqueous angiographic segmental patterns with bead-based methods which directly visualize segmental trabecular meshwork (TM) tracer trapping. Additionally, segmental protein expression differences between aqueous angiographic-derived low- and high-outflow human TM regions were evaluated. Post-mortem human eyes
Catia Morelli et al.
Oncogene, 23(45), 7517-7526 (2004-08-20)
Insulin receptor substrate 1 (IRS-1) is a major signaling molecule activated by the insulin and insulin-like growth factor I receptors. Recent data obtained in different cell models suggested that in addition to its conventional role as a cytoplasmic signal transducer
M G Myers et al.
Endocrinology, 132(4), 1421-1430 (1993-04-01)
IRS-1 is a unique cytosolic protein that becomes tyrosine phosphorylated during insulin stimulation of intact cells and immediately associates with the phosphatidylinositol 3'-kinase (PtdIns 3'-kinase). The insulin-like growth factor-I (IGF-I) receptor also mediated the tyrosine phosphorylation of IRS-1 and increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service