Skip to Content
Merck

Skip To

I0898

D-Ile-Pro-Arg p-nitroanilide dihydrochloride

≥95% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
CZK 13,900.00

About This Item

Empirical Formula (Hill Notation):
C23H36N8O5 · 2HCl
CAS Number:
Molecular Weight:
577.50
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:

CZK 13,900.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

solubility

water: 1 mg/mL, clear, colorless to faintly yellow

SMILES string

Cl.Cl.CC[C@@H](C)[C@@H](N)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C23H36N8O5.2ClH/c1-3-14(2)19(24)22(34)30-13-5-7-18(30)21(33)29-17(6-4-12-27-23(25)26)20(32)28-15-8-10-16(11-9-15)31(35)36;;/h8-11,14,17-19H,3-7,12-13,24H2,1-2H3,(H,28,32)(H,29,33)(H4,25,26,27);2*1H/t14-,17+,18+,19-;;/m1../s1

InChI key

NAANFMDBMZZFRD-CYTOJJGESA-N

General description

Chromogenic substrate for tissue plasminogen activator (t-PA).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S9009V7127C2276
assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (TLC)

solubility

water: 1 mg/mL, clear, colorless to faintly yellow

solubility

water: 25 mg/mL, clear, colorless to light yellow

solubility

water: 50 mg/mL, clear, light yellow

solubility

methanol: 20 mg/mL, clear, colorless to light yellow

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters