Key Documents
HS24322A
Mega Rack® tube rack
single, for 10-13 mm tubes, blue
Select a Size
1 EACH
CZK 1,330.00
CZK 1,330.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
CZK 1,330.00
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
CZK 1,330.00
Please contact Customer Service for Availability
material
polypropylene
feature
Holds 216 x 5-7 mL tubes
manufacturer/tradename
Heathrow Scientific HS24322A
size
337x235x91 mm
color
blue
General description
Mega Rack High-capacity storage rack for sample and blood collection tubes.
  • Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
  • Sturdy and light weight material for easy transport
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack and eliminate need for multiple tube racks
  • Autoclavable
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)