Skip to Content
Merck
All Photos(1)

Documents

HS234525B

Parafilm® Sealing Film Dispenser, Blue

blue (dispenser)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.51

material

acrylic
blue (dispenser)

feature

holds 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll

packaging

pack of 1 ea

manufacturer/tradename

Heathrow Scientific HS234525B

L × W × H

120 mm × 156 mm × 171 mm

Looking for similar products? Visit Product Comparison Guide

General description

The ABS dispenser has a unique design for maximum safety, storage, dispensing, and cutting Parafilm, tape, labels, and Tough-Tags.

Features and Benefits

  • ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to fingers.
  • Dispenser holds one or two 50 mm (2-inch) rolls, or one 100 mm (4-inch) roll
  • Supplied with two easily exchanged cutting blades

Legal Information

Parafilm is a registered trademark of Bemis Company, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service