Skip to Content
Merck
All Photos(1)

Key Documents

HS120521

Digital Orbital Shaker Replacement Silicon Mat and 4 Side Bumpers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
CZK 1,760.00

CZK 1,760.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
CZK 1,760.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.73

CZK 1,760.00


Please contact Customer Service for Availability

packaging

pkg of 1 ea

manufacturer/tradename

Heathrow Scientific 120521

General description

Silicon mat with side bumpers for digital orbital shaker.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, What is the dimension of this rubber sticky mat?

    1 answer
    1. The dimensions of this product are as follows:
      Length x Width: 11 x 11 inches
      Height: 0.32 inches

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service