Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
material
orange , white
feature
adjustable, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120100
volume measuring range
5-50 μL
General description
Features and Benefits
- Internal metal parts for increased accuracy and calibration stability, and long life
- Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
- Easy volume adjustment with locking tab to prevent accidental change
- Separate tip ejector
- Large digit volume display
- Color coded and labeled with volume range
- Both top and side have colored coded components for quick visual confirmation
- CE certified and Manufactured in accordance with ISO08655 and GLP regulations
- Autoclavable
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description adjustable, volume 5-50 μL, orange, single-channel | description adjustable, volume 5-50 μL, orange, 8-channel | description adjustable, volume 5-50 μL, orange, 12-channel | description adjustable, volume 0.1-2 μL, green, single-channel |
| material orange , white | material orange , white | material orange , white | material green , white |
| feature adjustable, single-channel | feature 8-channel, adjustable | feature 12-channel, adjustable | feature adjustable, single-channel |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| volume measuring range 5-50 μL | volume measuring range 5-50 μL | volume measuring range 5-50 μL | volume measuring range 0.1-2 μL |
| manufacturer/tradename Heathrow Scientific HS120100 | manufacturer/tradename Heathrow Scientific HS120215 | manufacturer/tradename Heathrow Scientific HS120217 | manufacturer/tradename Heathrow Scientific HS120097 |
Still not finding the right product?
Explore all of our products under Pearl™ Pipette
Choose from one of the most recent versions:
Already Own This Product?