Skip to Content
Merck
All Photos(6)

Documents

HPA026511

Sigma-Aldrich

Anti-CAMSAP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-CAMSAP1L1, Anti-KIAA1078

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TKGALSPITDNTEVDTGIHVPSEDIPETMDEDSSLRDYTVSLDSDMDDASKFLQDYDIRTGNTREALSPCPSTVSTKSQPGSSASSSSGVKM

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Calmodulin regulated spectrin associated protein family member 2 (CAMSAP2) is a cytoskeletal protein. The gene encoding it is localized on human chromosome 1q32.1.

Immunogen

Calmodulin-regulated spectrin-associated protein 1-like protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Calmodulin regulated spectrin associated protein family member 2 (CAMSAP2) associates with the minus-end of microtubules and plays a role in their organization in neurons. It also functions in dendrite branch formation, neuronal polarity and axon specification. CAMSAP2 may have a role in symptomatic epilepsy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76669

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Zhang et al.
Neuroscience letters, 556, 146-151 (2013-10-24)
In a recent genome-wide association study (GWAS) of symptomatic epilepsy in the Chinese population, the most significant single nucleotide polymorphism (SNP) allele was rs2292096 [G] (P=1.0×10(-8), odds ratio [OR]=0.63), in the CAMSAP1L1 gene (also known as CAMSAP2). Here, we report
Youling Guo et al.
Human molecular genetics, 21(5), 1184-1189 (2011-11-26)
In the majority of patients, epilepsy is a complex disorder with multiple susceptibility genes interacting with environmental factors. However, we understand little about its genetic risks. Here, we report the first genome-wide association study (GWAS) to identify common susceptibility variants
Kah Wai Yau et al.
Neuron, 82(5), 1058-1073 (2014-06-09)
In neurons, most microtubules are not associated with a central microtubule-organizing center (MTOC), and therefore, both the minus and plus-ends of these non-centrosomal microtubules are found throughout the cell. Microtubule plus-ends are well established as dynamic regulatory sites in numerous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service