Skip to Content
Merck
All Photos(3)

Key Documents

HPA019851

Sigma-Aldrich

Anti-TPMT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Thiopurine S-methyltransferase, Anti-Thiopurine methyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ALVAINPGDRKCYADTMFSLLGKKFQYLLCVLSYDPTKHPGPPFYVPHAEIERLFGKICNIRCLEKVDAFEERHKSWGIDCL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPMT(7172)

General description

TPMT (Thiopurine S-methyltransferase) is a cytoplasmic enzyme widely expressed in many cells. It is highly expressed in the liver and to a lower extent in the brain and lungs. It is mapped on the chromosome 6p22.3 region and has a molecular mass of ~35kDa.

Immunogen

Thiopurine S-methyltransferase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TPMT (Thiopurine S-methyltransferase) mainly mediates different catalytic S-methylation reaction such as 6-mercaptopurine and 6-thioguanine. It also participates in the metabolism of some drugs such as thiopurine drugs, which is largely influenced by TPMT polymorphisms. Deficiency in TPMT gene causes an autosomal recessive disorder,namely childhood acute lymphoblastic leukemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73837

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H L McLeod et al.
Leukemia, 14(4), 567-572 (2000-04-14)
Thiopurine methyltransferase (TPMT) catalyses the S-methylation of thiopurines, including 6-mercaptopurine and 6-thioguanine. TPMT activity exhibits genetic polymorphism, with about 1/300 inheriting TPMT deficiency as an autosomal recessive trait. If treated with standard doses of thiopurines, TPMTdeficient patients accumulate excessive thioguanine
Hyun-Young Kim et al.
Pharmacogenetics and genomics, 25(3), 143-146 (2015-01-08)
Thiopurine S-methyltransferase (TPMT) is a cytoplasmic enzyme involved in the metabolism of thiopurine drugs and its activity is largely influenced by polymorphisms of the TPMT gene. To date, more than 35 TPMT variants are known to be associated with reduced
Hamza Ben Zeglam et al.
The Libyan journal of medicine, 10, 27053-27053 (2015-03-31)
Thiopurine S-methyltransferase (TPMT) is a cytosolic enzyme that catalyses the S-methylation of 6-mercaptopurine and azathioprine. Low activity phenotypes are correlated with polymorphism in the TPMT gene. Patients with low or undetectable TMPT activity could develop severe myelosuppression when they are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service