Skip to Content
Merck
All Photos(2)

Key Documents

HPA015568

Sigma-Aldrich

Anti-LRRC3B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Leucine-rich repeat protein LRP15, Anti-Leucine-rich repeat-containing protein 3B precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

CPKGCLCSSSGGLNVTCSNANLKEIPRDLPPETVLLYLDSNQITSIPNEIFKDLHQLRVLNLSKNGIEFIDEHAFKGVAETLQTLDLSDNRIQSVHKNAFNNLKARARIANNPWHCDCTLQQVLRSMASNHETAHNVICKTSVLD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRRC3B(116135)

Immunogen

Leucine-rich repeat-containing protein 3B precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Leucine rich repeat containing 3B (LRRC3B) is mapped on human chromosome 3 in the 3p24 region. It is a potential tumor suppressor gene and may be involved in the process of carcinogenesis. This gene is associated with colon and gastric cancers and also in leukaemias. It may also act as a novel methylation-sensitive tumor suppressor gene in CRC (colorectal cancer). It may be a biomarker of CRC and breast cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72317

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Cell biochemistry and biophysics, 70(2), 1035-1041 (2014-05-20)
Epigenetic alteration of tumor suppressor genes by promoter hypermethylation has played a key role in tumorigenesis, which is an important mechanism as indispensable as gene deletion and mutation. LRRC3B is a potential tumor suppressor gene newly discovered; however, the specific
Xiao Qing Tian et al.
Scandinavian journal of gastroenterology, 44(1), 79-84 (2008-09-26)
Tumor suppressor gene silencing via promoter hypermethylation is an important event in the pathogenesis of colorectal cancer (CRC). Some aberrant DNA hypermethylation has high tumor specificity, so it may contribute to early diagnosis of CRC. The objective of this study
A G Kondratov et al.
Molecular medicine reports, 5(2), 509-512 (2011-11-22)
The leucine rich repeat containing 3B (LRRC3B) gene is a putative tumor suppressor located on human chromosome 3 in the 3p24 region. LRRC3B is frequently altered in colon and gastric cancers and also in leukaemias. In this study we investigated
Klas Haraldson et al.
Biochimie, 94(5), 1151-1157 (2012-02-11)
Chromosome 3 specific NotI microarrays containing 180 NotI linking clones associated with 188 genes were hybridized to NotI representation probes prepared using matched tumor/normal samples from major epithelial cancers: breast (47 pairs), lung (40 pairs) cervical (43 pairs), kidney (34
Gong-Sheng Li et al.
Oncology letters, 14(4), 5013-5017 (2017-11-01)
The present study aimed to investigate the potential effect of leucine-rich repeat containing 3B (LRRC3B) with respect to the inhibition of breast cancer recurrence and metastasis post-anesthesia. The mRNA expression of LRRC3B in breast MDA-MB-231 and MCF-7 cell lines was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service