Skip to Content
Merck
All Photos(2)

Key Documents

HPA011740

Sigma-Aldrich

Anti-FLT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Flt-1, Anti-Tyrosine-protein kinase FRT, Anti-Tyrosine-protein kinase receptor FLT, Anti-VEGFR-1, Anti-Vascular endothelial growth factor receptor 1, Anti-Vascular permeability factor receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable

immunogen sequence

LNLTIMNVSLQDSGTYACRARNVYTGEEILQKKEITIRDQEAPYLLRNLSDHTVAISSSTTLDCHANGVPEPQITWFKNNHKIQQEPGIILGPGSSTLFIERVTEEDEGVYHCKATNQKGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLT1(2321)

Immunogen

Vascular endothelial growth factor receptor 1 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-FLT1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FLT1 (Fms-related tyrosine kinase 1) is a tyrosine protein kinase with molecular mass of 180kDa. It acts as a scavenger receptor for the vascular endothelial growth factor (VEGF) in the proliferation of vascular endothelial cells. It is crucially important for promoting tumor growth, inflammation, epithelial to mesenchymal transition (EMT), regulation of monocyte migration, and enhances the adhesiveness of natural killer cells. Studies show that FLT1 may have a prognostic sensor activity in colorectal carcinoma (CRC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaudah Al-Maghrabi et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(9), 9045-9051 (2014-06-09)
Colorectal carcinoma (CRC) is a major cause of morbidity and mortality. Vascular endothelial growth factor 1/Fms-like tyrosine kinase 1 (VEGFR1/Flt-1) regulates monocyte migration, recruits endothelial cell progenitors, increases the adhesive properties of natural killer cells and induces of growth factors.
Evert Jan Van Limbergen et al.
Acta oncologica (Stockholm, Sweden), 53(5), 637-645 (2013-09-18)
Head and neck squamous cell carcinoma (HNSCC) is the fifth most common malignancy worldwide, responsible for approximately half a million new cases every year. The treatment of this disease is challenging and characterised by high rates of therapy failure and
L Seetharam et al.
Oncogene, 10(1), 135-147 (1995-01-05)
Flt-1 (fms-like tyrosine kinase-1), a receptor-type tyrosine kinase of sharing similar features with two other flt-family encoded proteins KDR/Flk-1 and Flt-4, has been recently identified as a receptor for Vascular Endothelial Growth Factor (VEGF) known to induce the proliferation of
John R Zibert et al.
The Journal of clinical investigation, 121(1), 410-421 (2010-12-08)
Dysregulated angiogenesis is a hallmark of chronic inflammatory diseases, including psoriasis, a common skin disorder that affects approximately 2% of the population. Studying both human psoriasis in 2 complementary xenotransplantation models and psoriasis-like skin lesions in transgenic mice with epidermal
Hui Wang et al.
Mutagenesis, 29(5), 311-317 (2014-06-04)
Lung inflammation and epithelial to mesenchymal transition (EMT) are two pathogenic features for the two contextual diseases: chronic obstructive pulmonary disease (COPD) and lung cancer. VEGFR1 (or FLT1) plays a certain role in promoting tumour growth, inflammation and EMT. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service