Skip to Content
Merck
All Photos(4)

Key Documents

HPA005513

Sigma-Aldrich

Anti-BIRC2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Baculoviral IAP repeat-containing protein 2, Anti-C-IAP1, Anti-HIAP-2, Anti-HIAP2, Anti-IAP homolog B, Anti-Inhibitor of apoptosis protein 2, Anti-RING finger protein 48, Anti-TNFR2-TRAF-signaling complex protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 12,900.00

CZK 12,900.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 12,900.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

CZK 12,900.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NWKLGDSPIQKHKQLYPSCSFIQNLVSASLGSTSKNTSPMRNSFAHSLSPTLEHSSLFSGSYSSLSPNPLNSRAVEDISSSRTNPYSYAMSTEEARFLTYHMWPLTFLSPSELARAGF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BIRC2(329)

General description

BIRC2 (baculoviral IAP repeat containing 2) is a member of the inhibitor of apoptosis protein (IAP) family. It is expressed almost exclusively in nucleus, which is mediated by its Bir domains. BIRC2 is a RING finger ubiquitin ligase (E3). This gene is localized to chromosome 11q22.

Immunogen

Baculoviral IAP repeat-containing protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Baculoviral IAP repeat containing 2 (BIRC2) is responsible for regulating NF-κB signaling, as well as apoptosis. It normally exists as a monomer, but is induced by proapoptotic signals to dimerize and undergo proteasomal degradation. It acts as an inhibitor of caspase, as well as a co-factor in tumor necrosis factor signaling. Studies suggest that, BIRC2 might regulate cell cycle, as it is accumulated in the midbody of cells in telophase stage. BIRC2 has also been proposed as an oncogene, as it degrades Mad-1, which is an antagonist of Myc. Degradation of Mad-1 leads to increased levels of Myc, which in turn promotes cell proliferation. It also regulates E2F1 transcription factor, and prevents its binding to DNA and E2F1-mediated transcriptional activation. BIRC2 is overexpressed in uterine cervix carcinoma of squamous cell type.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86198

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron H Phillips et al.
Nature structural & molecular biology, 21(12), 1068-1074 (2014-11-11)
Cellular inhibitor of apoptosis 1 (cIAP1) is a ubiquitin ligase with critical roles in the control of programmed cell death and NF-κB signaling. Under normal conditions, the protein exists as an autoinhibited monomer, but proapoptotic signals lead to its dimerization
Lei Xu et al.
Molecular cell, 28(5), 914-922 (2007-12-18)
c-IAP1, a member of the inhibitor of apoptosis protein (IAP) family and a RING finger ubiquitin ligase (E3), has been proposed to be an important oncogene. In many types of cancers, the levels of c-IAP1 are upregulated, which contributes positively
M Choschzick et al.
Virchows Archiv : an international journal of pathology, 461(2), 123-128 (2012-06-27)
Oncogene amplification is a key step in cell transformation towards malignancy. Chromosomal aberrations involving the long arm of chromosome 11, including amplifications at 11q13 and 11q22, have been previously reported in cervical cancer. While the role of the CCND1 gene
Konstantin Bagnjuk et al.
Journal of ovarian research, 12(1), 76-76 (2019-08-16)
Granulosa cell tumors (GCTs) are derived from proliferating granulosa cells of the ovarian follicle. They are known for their late recurrence and most patients with an aggressive form die from their disease. There are no treatment options for this slowly proliferating
Jessy Cartier et al.
The Journal of biological chemistry, 286(30), 26406-26417 (2011-06-10)
The inhibitor of apoptosis protein cIAP1 (cellular inhibitor of apoptosis protein-1) is a potent regulator of the tumor necrosis factor (TNF) receptor family and NF-κB signaling pathways in the cytoplasm. However, in some primary cells and tumor cell lines, cIAP1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service