Skip to Content
Merck
All Photos(4)

Key Documents

HPA004824

Sigma-Aldrich

Anti-H6PD antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-6PGL)] antibody produced in rabbit, Anti-GDH/6PGL endoplasmic bifunctional protein precursor [Includes: Glucose 1-dehydrogenase antibody produced in rabbit, Anti-Hexose-6-phosphate dehydrogenase); 6-phosphogluconolactonase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

WQGLFQLYLDEAGRGHSFSFHGAALTAPKQGQELMAKALESLSCPKDMAPSHCAEHKDQFLQLSQYRQLKTAEDYQALNKDIEAQLQHAGLREAGRIFYFSVPPFAYEDIAR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... H6PD(9563)

Looking for similar products? Visit Product Comparison Guide

Immunogen

GDH/6PGL endoplasmic bifunctional protein precursor [Includes: Glucose 1-dehydrogenase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

H6PD (hexose-6-phosphate dehydrogenase, glucose 1-dehydrogenase) is a microsomal enzyme involved in the regulation of peripheral corticosteroid metabolism in the liver and adipose tissue. It regenerates NADPH within the endoplasmic reticulum via the pentose phosphate pathway which is important for steroid hormone and drug metabolism. By supplying NADPH, it regulates the oxo-reductase activity of, 11-β hydroxysteroid dehydrogenase type I (11β-HSD1). The H6PD generated NADPH helps in reduction reaction of cortisone to cortisol in association with 11β-HSD1. Mutation in H6PD leads to the polycystic ovary syndrome (PCOS) and obesity with hyperandrogenemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86785

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Martínez-García et al.
Gene, 497(1), 38-44 (2012-02-07)
Hexose-6-phosphate dehydrogenase (H6PDH) influences 11β-hydroxysteroid dehydrogenase activity, a key enzyme in the peripheral metabolism of cortisol that modulates insulin sensitivity in adipose tissue. To study the associations of R453Q and D151A polymorphisms in the H6PDH gene (H6PD) with polycystic ovary
Thahira J Rahman et al.
PloS one, 6(8), e23248-e23248 (2011-08-23)
The combined thickness of the intima and media of the carotid artery (carotid intima-medial thickness, CIMT) is associated with cardiovascular disease and stroke. Previous studies indicate that carotid intima-medial thickness is a significantly heritable phenotype, but the responsible genes are
Michael Weingartner et al.
British journal of pharmacology, 178(16), 3309-3326 (2021-01-16)
11β-Hydroxysteroid dehydrogenase 1 (11β-HSD1) regulates tissue-specific glucocorticoid metabolism and its impaired expression and activity are associated with major diseases. Pharmacological inhibition of 11β-HSD1 is considered a promising therapeutic strategy. This study investigated whether alternative 7-oxo bile acid substrates of 11β-HSD1
Kenan Qin et al.
Steroids, 76(1-2), 135-139 (2010-11-06)
Hexose-6-phosphate dehydrogenase (H6PD) inactivating mutations cause cortisone reductase deficiency, which manifests with hyperandrogenism unexplained by commonly used tests and, thus, mimics polycystic ovary syndrome (PCOS). The aim of this study was to screen for mutations of H6PD gene in PCOS
Michael Weingartner et al.
PLoS neglected tropical diseases, 16(1), e0009192-e0009192 (2022-01-15)
Echinococcus multilocularis causes alveolar echinococcosis (AE), a rising zoonotic disease in the northern hemisphere. Treatment of this fatal disease is limited to chemotherapy using benzimidazoles and surgical intervention, with frequent disease recurrence in cases without radical surgery. Elucidating the molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service