Skip to Content
Merck
All Photos(1)

Key Documents

HPA004000

Sigma-Aldrich

Anti-ADAM9 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A disintegrin and metalloproteinase domain 9, Anti-ADAM 9 precursor, Anti-Cellular disintegrin-related protein, Anti-Meltrin-gamma, Anti-Metalloprotease/disintegrin/cysteine-rich protein 9, Anti-Myeloma cell metalloproteinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

PCQNNKAYCYNGMCQYYDAQCQVIFGSKAKAAPKDCFIEVNSKGDRFGNCGFSGNEYKKCATGNALCGKLQCENVQEIPVFGIVPAIIQTPSRGTKCWGVDFQLGSDVPDPGMVNEGTKCGAGKICRNFQCVDASVLNYDCD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM9(8754)

Immunogen

ADAM 9 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Disintegrin and metalloproteinase domain-containing protein 9 is an enzyme encoded by the ADAM9 gene in humans and is a member of the ADAM protein family, which contains a disintegrin domain. It is implicated in cancer cell migration and lung cancer metastasis to the brain. Its mRNA might be a potential biomarker for the differential diagnosis of pulmonary sarcoidosis. The protein plays a key role in many physiological processes, including fertilization, migration and cell survival. It is an important molecule in the processes of invasion and metastasis. It is up-regulated during cancer progression and hormone therapy.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86660.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingyu Liu et al.
The Journal of international medical research, 41(4), 1236-1241 (2013-07-17)
This prospective study investigated the potential role of serum A disintegrin and metalloproteinase 9 (ADAM9) mRNA in the differential diagnosis of pulmonary sarcoidosis from other diseases with similar clinical presentations, such as tuberculosis (TB). Serum samples were collected from patients
Kelli Cristina Micocci et al.
Biochimie, 95(7), 1371-1378 (2013-03-19)
ADAM9 (A Disintegrin And Metalloproteinase 9) is a member of the ADAM protein family which contains a disintegrin domain. This protein family plays key roles in many physiological processes, including fertilization, migration, and cell survival. The ADAM proteins have also
Che-Ming Liu et al.
PloS one, 8(1), e53795-e53795 (2013-01-24)
Cancer cells respond to stress by activating a variety of survival signaling pathways. A disintegrin and metalloproteinase (ADAM) 9 is upregulated during cancer progression and hormone therapy, functioning in part through an increase in reactive oxygen species. Here, we present
Chen-Yuan Lin et al.
Cancer research, 74(18), 5229-5243 (2014-07-26)
The transmembrane cell adhesion protein ADAM9 has been implicated in cancer cell migration and lung cancer metastasis to the brain, but the underpinning mechanisms are unclear and clinical support has been lacking. Here, we demonstrate that ADAM9 enhances the ability
Trang Thi-Huynh Le et al.
American journal of cancer research, 12(1), 176-197 (2022-02-11)
Metastatic and castration-resistant disease is a fatal manifestation of prostate cancer (PCa). The mechanism through which resistance to androgen deprivation in PCa is developed remains largely unknown. Our understanding of the tumor microenvironment (TME) and key signaling pathways between tumors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service