Skip to Content
Merck
All Photos(5)

Key Documents

HPA003404

Sigma-Aldrich

Anti-NDUFV2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial precursor antibody produced in rabbit, Anti-NADH-ubiquinone oxidoreductase 24 kDa subunit antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 14,200.00

CZK 14,200.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 14,200.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

CZK 14,200.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PEGHKAAAVLPVLDLAQRQNGWLPISAMNKVAEVLQVPPMRVYEVATFYTMYNRKPVGKYHIQVCTTTPCMLRNSDSILEAIQKKLGIKVGETTPDKLFTLIEVECLGACVNAPMVQINDNYYEDLTAKDIEE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDUFV2(4729)

Related Categories

Immunogen

NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-NDUFV2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

NDUFV2 (NADH dehydrogenase (ubiquinone) flavoprotein 2) gene encodes a 24kDa protein that forms a core subunit of NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain. This complex consists of 43 subunits and is involved in electron transfer from NADH to ubiquinone. The complex is localized to the inner mitochondrial membrane. Polymorphism in this gene may lead to a deficiency of complex I resulting in a higher risk of developing Parkinson disease. A deficiency in complex I resulting from a defect in this gene may cause hypertrophic cardiomyopathy and encephalopathy. Polymorphism in this gene may also cause bipolar disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86648

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Baertling et al.
Clinical genetics, 93(1), 111-118 (2017-07-04)
Mitochondrial respiratory chain complex I consists of 44 different subunits and contains 3 functional modules: the Q-, the N- and the P-module. NDUFA9 is a Q-module subunit required for complex I assembly or stability. However, its role in complex I
Paule Bénit et al.
Human mutation, 21(6), 582-586 (2003-05-20)
Respiratory chain complex I deficiencies represent a genetically heterogeneous group of diseases resulting from mutations in either mitochondrial or nuclear DNA. Combination of denaturing high performance liquid chromatography and sequence analysis allowed us to show that a 4-bp deletion in
Shinsuke Washizuka et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 120B(1), 72-78 (2003-06-20)
Linkage of bipolar disorder with 18p11 has been replicated by several investigators. A nuclear-encoded mitochondrial complex I subunit gene, NDUFV2, is one of the candidate genes in this locus, since the possible pathophysiological significance of mitochondrial dysfunction in bipolar disorder
N Hattori et al.
Genomics, 49(1), 52-58 (1998-05-08)
We analyzed the gene encoding the 24-kDa subunit of mitochondrial complex I, which has been implicated in the pathogenesis of Parkinson disease (PD). We set out to identify a polymorphism in the 24-kDa subunit gene (NDUFV2) in patients with PD
Xiu-Ming Wu et al.
Plant biotechnology journal, 14(9), 1925-1935 (2016-02-26)
Plant engineered to express double-stranded RNA (dsRNA) can target the herbivorous insect gene for silencing. Although mounting evidence has emerged to support feasibility of this new pest control technology, field application is slow largely due to lack of potent targets.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service